Tisk…

(4-Chlorophenyl)methyl acetate, tech

Dodavatel: Apollo Scientific

Synonyms: p-Chlorobenzyl acetate, 4-Chlorobenzyl acetate

OR32624-1G
APOSOR32624-1GEA 28630 CZK
APOSOR32624-1G
(4-Chlorophenyl)methyl acetate, tech
(4-Chlorophenyl)methyl acetate
Vzorec: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027406
CAS číslo: 5406-33-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR