Tisk…

2-(3,5-Dimethyl-4-propyl-1H-pyrazol-1-yl)pyridin-3-amine, tech

Dodavatel: Apollo Scientific

OR32586-1G
APOSOR32586-1GEA 44640 CZK
APOSOR32586-1G
2-(3,5-Dimethyl-4-propyl-1H-pyrazol-1-yl)pyridin-3-amine, tech
2-(3,5-Dimethyl-4-propyl-1H-pyrazol-1-yl)pyridin-3-amine
Vzorec: C₁₃H₁₈N₄
Storage Temperature: Ambient
MDL Number: MFCD03787559
CAS číslo: 955966-83-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR