Tisk…

4-Chloro-1,2-dimethoxybenzene 95%

Dodavatel: Apollo Scientific

OR32499-1G OR32499-5G
APOSOR32499-1GEA 2240 CZK
APOSOR32499-1G APOSOR32499-5G
4-Chloro-1,2-dimethoxybenzene 95%
4-Chloro-1,2-dimethoxybenzene
Vzorec: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD01741851
CAS číslo: 16766-27-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR