Tisk…

2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione, tech

Dodavatel: Apollo Scientific

OR32470-1G
APOSOR32470-1GEA 44640 CZK
APOSOR32470-1G
2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione, tech
2-Chloro-3-(pyrrolidin-1-yl)-1,4-dihydronaphthalene-1,4-dione
Vzorec: C₁₄H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD00010368
CAS číslo: 59641-25-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR