Tisk…

{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol, tech

Dodavatel: Apollo Scientific

OR32440-1G
APOSOR32440-1GEA 44640 CZK
APOSOR32440-1G
{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol, tech
{7-Methyl-2-phenylimidazo[1,2-a]pyridin-3-yl}methanol
Vzorec: C₁₅H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD01815092
CAS číslo: 217435-73-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR