Tisk…

5-Bromothiophene-2-carboxaldoxime

Dodavatel: Apollo Scientific

Synonyms: 5-Bromothiophene-2-carbaldehyde oxime

OR322321-500MG OR322321-1G
APOSOR322321-500MGEA 1450 CZK
APOSOR322321-500MG APOSOR322321-1G
5-Bromothiophene-2-carboxaldoxime
5-Bromothiophene-2-carboxaldoxime
Vzorec: C₅H₄BrNOS
MW: 206.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00126375
CAS číslo: 2160-63-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR