Tisk…

4-(N-Boc-aminomethyl)aniline

Dodavatel: Apollo Scientific

OR322111-5G OR322111-25G
APOSOR322111-5GEA 2070 CZK
APOSOR322111-5G APOSOR322111-25G
4-(N-Boc-aminomethyl)aniline
4-(N-Boc-aminomethyl)aniline
Vzorec: C₁₂H₁₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03001716
CAS číslo: 94838-55-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR