Tisk…

5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole

Dodavatel: Apollo Scientific

OR32162-1G
APOSOR32162-1GEA 6220 CZK
APOSOR32162-1G
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole
Vzorec: C₁₂H₁₀N₄
Storage Temperature: Ambient
MDL Number: MFCD00138185
CAS číslo: 321385-58-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR