Tisk…

1-(tert-Butoxycarbonyl)azepane-2-carboxylic acid

Dodavatel: Apollo Scientific

OR321523-1G OR321523-250MG
APOSOR321523-1GEA 13230 CZK
APOSOR321523-1G APOSOR321523-250MG
1-(tert-Butoxycarbonyl)azepane-2-carboxylic acid
1-(tert-Butoxycarbonyl)azepane-2-carboxylic acid
Vzorec: C₁₂H₂₁NO₄
MW: 243.299 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03790979
CAS číslo: 1034708-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR