Tisk…

(3Z)-3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech

Dodavatel: Apollo Scientific

OR32142-1G
APOSOR32142-1GEA 44640 CZK
APOSOR32142-1G
(3Z)-3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methylidene]-2,3-dihydro-1H-indol-2-one, tech
(3Z)-3-[(4-Hydroxy-3-methoxy-5-methylphenyl)methylidene]-2,3-dihydro-1H-indol-2-one
Vzorec: C₁₇H₁₅NO₃
MW: 281,306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05668727
CAS číslo: 860648-89-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR