Tisk…

tert-Butyl (1-amino-2-oxoazetidin-3-yl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

OR321290-250MG
APOSOR321290-250MGEA 32240 CZK
APOSOR321290-250MG
tert-Butyl (1-amino-2-oxoazetidin-3-yl)carbamate 95%
tert-Butyl (1-amino-2-oxoazetidin-3-yl)carbamate
Vzorec: C₈H₁₅N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD24539163
CAS číslo: 1822581-31-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR