Tisk…

5-Bromo-2-hydroxy-3-picoline

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Bromo-3-methylpyridin-2-ol, 5-Bromo-2-hydroxy-3-methylpyridine, 5-Bromo-3-methylpyridin-2(1H)-one

OR3202-5G
APOSOR3202-5GEA 622 CZK
APOSOR3202-5G
5-Bromo-2-hydroxy-3-picoline
5-Bromo-2-hydroxy-3-picoline
Vzorec: C₆H₆BrNO
MW: 188.02 g/mol
Bod tání: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD03427657
CAS číslo: 89488-30-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR