Tisk…

Ethyl-2-(3-iodophenoxy)acetate 95+%

Dodavatel: Apollo Scientific

OR318049-1G OR318049-10G OR318049-5G
APOSOR318049-1GEA 7770 CZK
APOSOR318049-1G APOSOR318049-10G APOSOR318049-5G
Ethyl-2-(3-iodophenoxy)acetate 95+%
Ethyl-2-(3-iodophenoxy)acetate
Vzorec: C₁₀H₁₁IO₃
Storage Temperature: Ambient
MDL Number: MFCD06659701
CAS číslo: 90888-04-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR