Tisk…

N-Boc-D-phenylalaninol

Dodavatel: Apollo Scientific

OR316042-100G OR316042-25G OR316042-5G
APOSOR316042-100GEA 6290 CZK
APOSOR316042-100G APOSOR316042-25G APOSOR316042-5G
N-Boc-D-phenylalaninol
N-Boc-D-phenylalaninol
Vzorec: C₁₄H₂₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00216472
CAS číslo: 106454-69-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR