Tisk…

Ethyl-1-cyanocyclopropanecarboxylate 96%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 1-Cyano-1-cyclopropanecarboxylate

OR315517-100G
APOSOR315517-100GEA 3970 CZK
APOSOR315517-100G
Ethyl-1-cyanocyclopropanecarboxylate 96%
Ethyl-1-cyanocyclopropanecarboxylate
Vzorec: C₇H₉NO₂
MW: 139.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00040159
CAS číslo: 1558-81-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR