Tisk…

8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane 97%

Dodavatel: Apollo Scientific

OR312319-250MG OR312319-100MG
APOSOR312319-250MGEA 16040 CZK
APOSOR312319-250MG APOSOR312319-100MG
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane 97%
8-Boc-2-oxa-5,8-diazaspiro[3.5]nonane
Vzorec: C₁₁H₂₀N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22035557
CAS číslo: 1367777-12-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR