Tisk…

Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride

Dodavatel: Apollo Scientific
Varování

OR312113-1G OR312113-250MG
APOSOR312113-1GEA 27540 CZK
APOSOR312113-1G APOSOR312113-250MG
Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride
Bicyclo[1.1.1]pentane-1,3-diamine dihydrochloride
Vzorec: C₅H₁₀N₂·2ClH
Storage Temperature: Ambient
MDL Number: MFCD19211809
CAS číslo: 147927-61-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR