Tisk…

Methyl-3-[(methylamino)sulphonyl]-1-benzothiophene-2-carboxylate

Dodavatel: Apollo Scientific

OR310781-1G
APOSOR310781-1GEA 22220 CZK
APOSOR310781-1G
Methyl-3-[(methylamino)sulphonyl]-1-benzothiophene-2-carboxylate
Methyl-3-[(methylamino)sulphonyl]-1-benzothiophene-2-carboxylate
Vzorec: C₁₁H₁₁NO₄S₂
MW: 285,34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105040
CAS číslo: 1325304-23-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR