Tisk…

5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]

Dodavatel: Apollo Scientific

OR310531-1G
APOSOR310531-1GEA 30290 CZK
APOSOR310531-1G
5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]
5,5-Dimethyl-3',3'a,4',6'a-tetrahydro-1'H-spiro[1,3-dioxane-2,2'-pentalene]
Vzorec: C₁₃H₂₀O₂
MW: 208.297 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27957338
CAS číslo: 92007-41-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR