Tisk…

Methyl 2-(4-iodophenoxy)acetate

Dodavatel: Apollo Scientific

OR310454-5G
APOSOR310454-5GEA 6780 CZK
APOSOR310454-5G
Methyl 2-(4-iodophenoxy)acetate
Methyl 2-(4-iodophenoxy)acetate
Vzorec: C₉H₉IO₃
MW: 292,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07113288
CAS číslo: 81720-18-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR