Tisk…

7-Ethylquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific

OR309226-1G
APOSOR309226-1GEA 14680 CZK
APOSOR309226-1G
7-Ethylquinoline-3-carboxylic acid
7-Ethylquinoline-3-carboxylic acid
Vzorec: C₁₂H₁₁NO₂
MW: 201.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09787862
CAS číslo: 948290-70-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR