Tisk…

3-Methyl-3-oxetanemethanol

Dodavatel: Apollo Scientific
Varování

OR309068-100G OR309068-25G OR309068-500G
APOSOR309068-100GEA 1370 CZK
APOSOR309068-100G APOSOR309068-25G APOSOR309068-500G
3-Methyl-3-oxetanemethanol
3-Methyl-3-oxetanemethanol
Vzorec: C₅H₁₀O₂
Bod varu: 78…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010273
CAS číslo: 3143-02-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR