Tisk…

Methyl (2S)-morpholine-2-carboxylate hydrochloride

Dodavatel: Apollo Scientific

OR308164-1G
APOSOR308164-1GEA 2780 CZK
APOSOR308164-1G
Methyl (2S)-morpholine-2-carboxylate hydrochloride
Methyl (2S)-morpholine-2-carboxylate hydrochloride
Vzorec: C₆H₁₁NO₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD21648490
CAS číslo: 1417789-45-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR