Tisk…

1-Boc-4-amino-4-methylpiperidine

Dodavatel: Apollo Scientific

OR308122-1G OR308122-5G
APOSOR308122-1GEA 2610 CZK
APOSOR308122-1G APOSOR308122-5G
1-Boc-4-amino-4-methylpiperidine
1-Boc-4-amino-4-methylpiperidine
Vzorec: C₁₁H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03265497
CAS číslo: 343788-69-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR