Tisk…

4-Phenyl-tetrahydro-pyran-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR30811-1G OR30811-250MG
APOSOR30811-1GEA 3310 CZK
APOSOR30811-1G APOSOR30811-250MG
4-Phenyl-tetrahydro-pyran-4-carboxylic acid
4-Phenyl-tetrahydro-pyran-4-carboxylic acid
Vzorec: C₁₂H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00085758
CAS číslo: 182491-21-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR