Tisk…

Cyclobutylmethanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

OR308087-1G OR308087-5G
APOSOR308087-1GEA 622 CZK
APOSOR308087-1G APOSOR308087-5G
Cyclobutylmethanamine hydrochloride
Cyclobutylmethanamine hydrochloride
Vzorec: C₅H₁₂ClN
MW: 121.6100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034952
CAS číslo: 5454-82-0

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR