Tisk…

tert-Butyl-4-aminobutyl(methyl)carbamate

Dodavatel: Apollo Scientific
Varování

OR308002-1G
APOSOR308002-1GEA 4100 CZK
APOSOR308002-1G
tert-Butyl-4-aminobutyl(methyl)carbamate
tert-Butyl-4-aminobutyl(methyl)carbamate
Vzorec: C₁₀H₂₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03001681
CAS číslo: 144222-23-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR