Tisk…

4-Amino-8-bromoquinoline

Dodavatel: Apollo Scientific

OR307955-1G OR307955-5G OR307955-250MG
APOSOR307955-1GEA 13060 CZK
APOSOR307955-1G APOSOR307955-5G APOSOR307955-250MG
4-Amino-8-bromoquinoline
4-Amino-8-bromoquinoline
Vzorec: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD04037193
CAS číslo: 65340-75-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR