Tisk…

6-Bromo-4-chloro-2-ethylquinoline

Dodavatel: Apollo Scientific

OR307872-1G
APOSOR307872-1GEA 15640 CZK
APOSOR307872-1G
6-Bromo-4-chloro-2-ethylquinoline
6-Bromo-4-chloro-2-ethylquinoline
Vzorec: C₁₁H₉BrClN
Storage Temperature: Ambient
MDL Number: MFCD09260433
CAS číslo: 930570-40-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR