Tisk…

3-(2-Methoxybenzyl)piperidine

Dodavatel: Apollo Scientific

OR307728-1G
APOSOR307728-1GEA 10680 CZK
APOSOR307728-1G
3-(2-Methoxybenzyl)piperidine
3-(2-Methoxybenzyl)piperidine
Vzorec: C₁₃H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD05863594
CAS číslo: 420137-10-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR