Tisk…

trans-2-(4-Methoxy-benzylamino)-cyclohexanol

Dodavatel: Apollo Scientific

OR307636-1G
APOSOR307636-1GEA 15440 CZK
APOSOR307636-1G
trans-2-(4-Methoxy-benzylamino)-cyclohexanol
trans-2-(4-Methoxy-benzylamino)-cyclohexanol
Vzorec: C₁₄H₂₁NO₂
MW: 235,322 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366628
CAS číslo: 759402-65-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR