Tisk…

1,2,3,4-Tetrahydroisoquinoline-1-acetic acid

Dodavatel: Apollo Scientific

OR307541-1G
APOSOR307541-1GEA 9030 CZK
APOSOR307541-1G
1,2,3,4-Tetrahydroisoquinoline-1-acetic acid
1,2,3,4-Tetrahydroisoquinoline-1-acetic acid
Vzorec: C₁₁H₁₃NO₂
MW: 191.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00215024
CAS číslo: 105400-81-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR