Tisk…

Methyl-1-methyl-1H-indazole-4-carboxylate

Dodavatel: Apollo Scientific
Varování

OR30718-1G OR30718-250MG
APOSOR30718-1GEA 953 CZK
APOSOR30718-1G APOSOR30718-250MG
Methyl-1-methyl-1H-indazole-4-carboxylate
Methyl-1-methyl-1H-indazole-4-carboxylate
Vzorec: C₁₀H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11109402
CAS číslo: 1071428-42-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR