Tisk…

(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95+%

Dodavatel: Apollo Scientific

OR307094-5G
APOSOR307094-5GEA 2860 CZK
APOSOR307094-5G
(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95+%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride
Vzorec: C₄H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD03840366
CAS číslo: 336100-47-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR