Tisk…

Fmoc-Hyp-OH 98%

Dodavatel: Apollo Scientific

OR307070-100G OR307070-25G
APOSOR307070-100GEA 5890 CZK
APOSOR307070-100G APOSOR307070-25G
Fmoc-Hyp-OH 98%
Fmoc-Hyp-OH
Vzorec: C₂₀H₁₉NO₅
Bod tání: 189…193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151929
CAS číslo: 88050-17-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR