Tisk…

Bis(triphenylphosphine)palladium(II) chloride 98%

Dodavatel: Apollo Scientific
Varování

OR30692-1G OR30692-25G OR30692-5G
APOSOR30692-1GEA 1160 CZK
APOSOR30692-1G APOSOR30692-25G APOSOR30692-5G
Bis(triphenylphosphine)palladium(II) chloride 98%
Bis(triphenylphosphine)palladium(II) chloride
Vzorec: C₃₆H₃₀Cl₂P₂Pd
Storage Temperature: Refrigerator
MDL Number: MFCD00009593
CAS číslo: 13965-03-2
EINECS: 237-744-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR