Tisk…

5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%

Dodavatel: Apollo Scientific

Synonyms: 5-Bromo-1H-pyrrolo[2,3-b]pyridin-2-one

OR305356-5G
APOSOR305356-5GEA 2320 CZK
APOSOR305356-5G
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one 97%
5-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Vzorec: C₇H₅BrN₂O
MW: 213.0300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367221
CAS číslo: 183208-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR