Tisk…

N-Boc-5-hydroxyindole

Dodavatel: Apollo Scientific

OR305227-1G OR305227-250MG
APOSOR305227-1GEA 2900 CZK
APOSOR305227-1G APOSOR305227-250MG
N-Boc-5-hydroxyindole
N-Boc-5-hydroxyindole
Vzorec: C₁₃H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD08272246
CAS číslo: 434958-85-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR