Tisk…

Imidazo[1,2-a]pyridine-3-sulphonyl chloride

Dodavatel: Apollo Scientific

OR305155-250MG OR305155-1G OR305155-500MG
APOSOR305155-250MGEA 10060 CZK
APOSOR305155-250MG APOSOR305155-1G APOSOR305155-500MG
Imidazo[1,2-a]pyridine-3-sulphonyl chloride
Imidazo[1,2-a]pyridine-3-sulphonyl chloride
Vzorec: C₇H₅ClN₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD03407340
CAS číslo: 499770-78-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR