Tisk…

3-Methoxy-2-methylbenzeneboronic acid 95%

Dodavatel: Apollo Scientific
Varování

OR305006-1G
APOSOR305006-1GEA 4570 CZK
APOSOR305006-1G
3-Methoxy-2-methylbenzeneboronic acid 95%
3-Methoxy-2-methylbenzeneboronic acid
Vzorec: C₈H₁₁BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18398348
CAS číslo: 1313617-76-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR