Tisk…

Chloromaleic acid anhydride

Dodavatel: Apollo Scientific
Nebezpečí

OR30483-1G OR30483-250MG
APOSOR30483-1GEA 0 CZK
APOSOR30483-1G APOSOR30483-250MG
Chloromaleic acid anhydride
Chloromaleic acid anhydride
Vzorec: C₄HClO₃
MW: 132.502 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210253
CAS číslo: 96-02-6
UN: 2923
ADR: 8+6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR