Tisk…

2-Methyl-2-(phenylamino)propionitrile

Dodavatel: Apollo Scientific

OR303950-1G OR303950-500MG
APOSOR303950-1GEA 7670 CZK
APOSOR303950-1G APOSOR303950-500MG
2-Methyl-2-(phenylamino)propionitrile
2-Methyl-2-(phenylamino)propionitrile
Vzorec: C₁₀H₁₂N₂
MW: 160.22 g/mol
Bod tání: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00035943
CAS číslo: 2182-38-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR