Tisk…

6-(Methylsulphonyl)nicotinaldehyde

Dodavatel: Apollo Scientific

OR303790-500MG
APOSOR303790-500MGEA 9560 CZK
APOSOR303790-500MG
6-(Methylsulphonyl)nicotinaldehyde
6-(Methylsulphonyl)nicotinaldehyde
Vzorec: C₇H₇NO₃S
MW: 185.2 g/mol
Bod tání: 114…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16140366
CAS číslo: 1221792-08-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR