Tisk…

(2,4-Dimethylthiazol-5-yl)acetic acid

Dodavatel: Apollo Scientific

OR303204-1G OR303204-500MG
APOSOR303204-1GEA 9560 CZK
APOSOR303204-1G APOSOR303204-500MG
(2,4-Dimethylthiazol-5-yl)acetic acid
(2,4-Dimethylthiazol-5-yl)acetic acid
Vzorec: C₇H₉NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01681584
CAS číslo: 34272-65-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR