Tisk…

3,5-Dibromophenoxytrimethylsilane

Dodavatel: Apollo Scientific

OR303200-1G OR303200-500MG
APOSOR303200-1GEA 7670 CZK
APOSOR303200-1G APOSOR303200-500MG
3,5-Dibromophenoxytrimethylsilane
3,5-Dibromophenoxytrimethylsilane
Vzorec: C₉H₁₂Br₂OSi
MW: 324.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23136036
CAS číslo: 856766-98-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR