Tisk…

(R)-2-Methoxypropanoic acid

Dodavatel: Apollo Scientific

OR302445-1G
APOSOR302445-1GEA 2360 CZK
APOSOR302445-1G
(R)-2-Methoxypropanoic acid
(R)-2-Methoxypropanoic acid
Vzorec: C₄H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD01632582
CAS číslo: 23943-96-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR