Tisk…

5-(Pyridin-2-yl)-1H-pyrazol-4-amine

Dodavatel: Apollo Scientific

OR302335-100MG OR302335-250MG OR302335-500MG
APOSOR302335-100MGEA 15710 CZK
APOSOR302335-100MG APOSOR302335-250MG APOSOR302335-500MG
5-(Pyridin-2-yl)-1H-pyrazol-4-amine
5-(Pyridin-2-yl)-1H-pyrazol-4-amine
Vzorec: C₈H₈N₄
Storage Temperature: Ambient
MDL Number: MFCD22690777
CAS číslo: 896467-81-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR