Tisk…

tert-Butyl-5-bromoisoindoline-2-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 5-bromoisoindoline-2-carboxylate

OR302276-5G
APOSOR302276-5GEA 1870 CZK
APOSOR302276-5G
tert-Butyl-5-bromoisoindoline-2-carboxylate
tert-Butyl-5-bromoisoindoline-2-carboxylate
Vzorec: C₁₃H₁₆BrNO₂
MW: 298.1800 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953049
CAS číslo: 201940-08-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR