Tisk…

1-(2-Methoxypyridin-3-yl)ethanone

Dodavatel: Apollo Scientific

OR302200-5G
APOSOR302200-5GEA 3610 CZK
APOSOR302200-5G
1-(2-Methoxypyridin-3-yl)ethanone
1-(2-Methoxypyridin-3-yl)ethanone
Vzorec: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847670
CAS číslo: 131674-40-3

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR