Tisk…

Methyl 4-(methylsulphonyl)benzoate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 4-(Methylsulphonyl)benzoate

OR301368-5G
APOSOR301368-5GEA 3940 CZK
APOSOR301368-5G
Methyl 4-(methylsulphonyl)benzoate 98%
Methyl 4-(methylsulphonyl)benzoate
Vzorec: C₉H₁₀O₄S
MW: 214.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00100037
CAS číslo: 22821-70-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR